Learn More
New England Biolabs, Inc. Terminal Transferase – 2500 units
SDP
Catalog No. 50995238
Encompass
Terminal transferase (TdT) is a template independent polymerase that catalyzes the addition of deoxynucleotides to the 3' hydroxyl terminus of DNA molecules. Protruding, recessed or blunt-ended double or single-stranded DNA molecules serve as a substrate for TdT. The 58.3 kDa enzyme does not have 5' or 3' exonuclease activity. The addition of Co2+ in the reacton makes tailing more efficient.
Catalog No. 50-995-238 Supplier New England Biolabs, Inc. Supplier No. M0315L
Terminal transferase (TdT) is a template independent polymerase that catalyzes the addition of deoxynucleotides to the 3' hydroxyl terminus of DNA molecules. Protruding, recessed or blunt-ended double or single-stranded DNA molecules serve as a substrate for TdT. The 58.3 kDa enzyme does not have 5' or 3' exonuclease activity. The addition of Co2+ in the reacton makes tailing more efficient.