Learn More
McMaster-Carr TERMINAL BLOCK GRAY ENDCVR
Supplier:  McMaster-Carr 7641K72
Encompass
McMaster-Carr offers a wide selection of terminal blocks for electrical connections. Their terminal blocks come in various sizes, materials, number of poles, and mounting styles to suit different applications. The blocks allow wires to be securely connected and provide a reliable electrical connection point. Key features include screw, spring, and push-in style wire terminations, ratings up to 600V and 63A, and materials like nylon, polyamide, and phenolic. The blocks can be panel, rail, or surface mounted. This versatile product line enables creating customized electrical connections for industrial, commercial, and residential projects.
Catalog No. NC1457115