Learn More
Qiagen Beverly Inc TERMINAL DEOXY TRANSFERASE
Catalog No. 50305927
Encompass
Terminal Deoxy Transferase (TdT); Conc. 20,000 U/ul; 6,000 U; incl. 10X buffer
Catalog No. 50-305-927 Supplier Qiagen Beverly Inc Supplier No. P7070L
Terminal Deoxy Transferase (TdT); Conc. 20,000 U/ul; 6,000 U; incl. 10X buffer