Learn More
Sigma Aldrich Fine Chemicals Biosciences Terminal Transferase from
Catalog No. 502905237
Encompass
Terminal Transferase catalyzes the template independent addition of deoxy- and dideoxynucleoside triphosphates to the 3-OH ends of double and single-stranded DNA fragments and oligonucleotides. Terminal Transferase incorporates digoxigenin- biotin- and fluorochrome-labeled deoxy- and dideoxynucleoside triphosphates as well as radioactively labeled deoxy- and dideoxynucleoside triphosphates. The supplied 5x-concentrated reaction buffer allows the optimal tailing of all types of double-stranded DNA ends blunt ended with 3 overhang or with 5 overhang. The highest incorporation rates are obtained with 3 overhangs.
Catalog No. 50-290-5237 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3333574001
Terminal Transferase catalyzes the template independent addition of deoxy- and dideoxynucleoside triphosphates to the 3-OH ends of double and single-stranded DNA fragments and oligonucleotides. Terminal Transferase incorporates digoxigenin- biotin- and fluorochrome-labeled deoxy- and dideoxynucleoside triphosphates as well as radioactively labeled deoxy- and dideoxynucleoside triphosphates. The supplied 5x-concentrated reaction buffer allows the optimal tailing of all types of double-stranded DNA ends blunt ended with 3 overhang or with 5 overhang. The highest incorporation rates are obtained with 3 overhangs.