Learn More
Cayman Chemical Termitomycamide B, Purity: ≥97%, CAS: 1254277-89-8, 10 mg
SDP
Catalog No. 502954057
Encompass_Preferred
A fatty acid amide from T. titanicus that at 0.1 μg/ml protects against ER stress-dependent cell death in Neuro2a cells induced by tunicamycin
Catalog No. 50-295-4057 Supplier Cayman Chemical Supplier No. 1119010 MG
A fatty acid amide from T. titanicus that at 0.1 μg/ml protects against ER stress-dependent cell death in Neuro2a cells induced by tunicamycin