Learn More
Cayman Chemical TermItomycamIde E 10mg
SDP
Catalog No. 502954058
Encompass_Preferred
A fatty acid amide from T. titanicus that at 0.1 μg/ml protects against ER stress-dependent cell death in Neuro2a cells induced by tunicamycin
Catalog No. 50-295-4058 Supplier Cayman Chemical Supplier No. 1119110 MG
A fatty acid amide from T. titanicus that at 0.1 μg/ml protects against ER stress-dependent cell death in Neuro2a cells induced by tunicamycin