Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ternidazole (hydrochloride) | 70028-95-4 | 98.55% | 221.64 | 1 ML
SDP

Catalog No. 5000351169
Encompass_Preferred

Ternidazole (hydrochloride) is a hydroxymetabolite of nitroimidazole with antiprotozoic properties, intended for research use only. It is utilized as a laboratory chemical and in the manufacture of substances.

  • Off-white to light yellow solid
  • Hydroxymetabolite of nitroimidazole
  • Exhibits antiprotozoic properties
  • Soluble in DMSO (100 mg/mL)
  • Soluble in H2O (100 mg/mL)
  • Purity (LCMS) of 98.55%

Catalog No. 50-003-51169 Supplier Medchemexpress LLC Supplier No. HY13643610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ternidazole (hydrochloride) is a hydroxymetabolite of nitroimidazole with antiprotozoic properties, intended for research use only. It is utilized as a laboratory chemical and in the manufacture of substances.

  • Off-white to light yellow solid
  • Hydroxymetabolite of nitroimidazole
  • Exhibits antiprotozoic properties
  • Soluble in DMSO (100 mg/mL)
  • Soluble in H2O (100 mg/mL)
  • Purity (LCMS) of 98.55%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.