Learn More
AdipoGen Terpendole C (1 mg)
SDP
Catalog No. 502053496
Encompass_Preferred
Chemical. CAS: 156967-65-6. Formula: C32H41NO5. MW: 519.7. Isolated from Albophoma yamanashiensis. Acyl-CoA:cholesterol acyltransferase (ACAT) isozymes ACAT1 and ACAT2 inhibitor. Tremorgenic. Cholesteryl ester (CE) synthesis inhibitor.
Catalog No. 50-205-3496 Supplier AdipoGen Supplier No. AGCN20125M001
Chemical. CAS: 156967-65-6. Formula: C32H41NO5. MW: 519.7. Isolated from Albophoma yamanashiensis. Acyl-CoA:cholesterol acyltransferase (ACAT) isozymes ACAT1 and ACAT2 inhibitor. Tremorgenic. Cholesteryl ester (CE) synthesis inhibitor.