Learn More
AdipoGen Terpendole E (250 µg)
SDP
Supplier:  AdipoGen AGCN20127C250
Encompass_Preferred
Chemical. CAS: 167427-23-8. Formula: C28H39NO3. MW: 437.6. Isolated from Albophoma yamanashiensis. Acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor. Mitotic kinesin Eg5 (Mitotic Kinesin Spindle Protein; KSP) inhibitor. Specific M phase inhibitor.
Catalog No. 50-205-3499