Learn More
Medchemexpress LLC HY-119821 1mg Medchemexpress, Terphenyllin CAS:52452-60-5 Purity:>98%
SDP
Catalog No. 501932542
Encompass_Preferred
Medchemexpress, HY-119821 1mg Terphenyllin CAS:52452-60-5 Terphenyllin is a naturally abundant p-terphenyl metabolite isolated from the coral derived fungus Aspergillus candidus, has significant α-glucosidase inhibitory activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2542 Supplier Medchemexpress LLC Supplier No. HY1198211MG
Medchemexpress, HY-119821 1mg Terphenyllin CAS:52452-60-5 Terphenyllin is a naturally abundant p-terphenyl metabolite isolated from the coral derived fungus Aspergillus candidus, has significant α-glucosidase inhibitory activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.