Learn More
TARGETMOL CHEMICALS INC TERPHENYLLIN 5MG
SDP
Catalog No. 502581379
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Terphenyllin has (alpha)-glucosidase inhibitory activity and is a natural product isolated from the coral derived fungus Aspergillus candidus. purity: 96%
Catalog No. 50-258-1379 Supplier TARGETMOL CHEMICALS INC Supplier No. T131305MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Terphenyllin has (alpha)-glucosidase inhibitory activity and is a natural product isolated from the coral derived fungus Aspergillus candidus. purity: 96%