Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terpin Hydrate, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607296 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
45 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-296 2.5 kg Poly Pail
18-607-297 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-296 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TE1102.5KG
Add to Cart
Add to Cart

TE110, 2451-01-6, C10H20O2·H2O

Spectrum™ Chemical Terpin Hydrate, USP is used to control a cough associated with a cold or other respiratory illnesses. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2451-01-6
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
InChI Key JGKJMBOJWVAMIJ-UHFFFAOYSA-N
IUPAC Name 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol hydrate
SMILES O.CC(C)(O)C1CCC(C)(O)CC1

Specifications

CAS Max % 1
Color White
Ignition Residue 0.001
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 98 to 100.5%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material Terpin Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.