Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terpin Monohydrate 98.0+%, TCI America™
SDP

Catalog No. T234425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T234425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T234425G Supplier TCI America Supplier No. T234425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2451-01-6
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
MDL Number MFCD00149225
InChI Key JGKJMBOJWVAMIJ-UHFFFAOYSA-N
Synonym terpin hydrate, p-menthane-1,8-diol monohydrate, terpin monohydrate, terpin cis-form hydrate, 4-2-hydroxypropan-2-yl-1-methylcyclohexanol hydrate, terpin hydrate usp, terpin hydrate cis-form, cyclohexanemethanol, 4-hydroxy-alpha,alpha,4-trimethyl-, monohydrate
PubChem CID 17141
IUPAC Name 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol;hydrate
SMILES CC1(CCC(CC1)C(C)(C)O)O.O

Specifications

Melting Point 119°C
Color White
Quantity 25 g
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Terpin Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.