Learn More
Medchemexpress LLC HY-W017316 100mg Medchemexpress, Terpinen-4-ol CAS:562-74-3 Purity:>98%
SDP
Catalog No. 501970726
Encompass_Preferred
Medchemexpress, HY-W017316 100mg Terpinen-4-ol CAS:562-74-3 Terpinen-4-ol (4-Carvomenthenol), a naturally occurring monoterpene, is the main bioactive component of tea-tree oil. Terpinen-4-ol suppresses inflammatory mediator production by activated human monocytes. Terpinen-4-ol significantly enhances the effect of several chemotherapeutic and biological agents[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0726 Supplier Medchemexpress LLC Supplier No. HYW017316100MG
Medchemexpress, HY-W017316 100mg Terpinen-4-ol CAS:562-74-3 Terpinen-4-ol (4-Carvomenthenol), a naturally occurring monoterpene, is the main bioactive component of tea-tree oil. Terpinen-4-ol suppresses inflammatory mediator production by activated human monocytes. Terpinen-4-ol significantly enhances the effect of several chemotherapeutic and biological agents[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.