Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-Terpinen-4-ol | 20126-76-5 | 1 G
SDP

Catalog No. 5000419113
Encompass_Preferred

(-)-Terpinen-4-ol is a monoterpene alcohol that can be extracted from Bark Beetle Polygraphus poligraphus. It has a molecular weight of 154.25 and a formula of C10H18O. This liquid appears colorless to light yellow and is derived from Plants Theaceae Camellia sinensis (L.) O. Ktze. It is intended for research use only.

  • Appearance: colorless to light yellow liquid
  • Molecular weight: 154.25
  • Molecular formula: C10H18O
  • Purity: 98.49%
  • Density: 0.933±0.06 g/cm3
  • For research use only
  • Extracted from natural sources

Catalog No. 50-004-19113 Supplier Medchemexpress LLC Supplier No. HYN79271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Terpinen-4-ol is a monoterpene alcohol that can be extracted from Bark Beetle Polygraphus poligraphus. It has a molecular weight of 154.25 and a formula of C10H18O. This liquid appears colorless to light yellow and is derived from Plants Theaceae Camellia sinensis (L.) O. Ktze. It is intended for research use only.

  • Appearance: colorless to light yellow liquid
  • Molecular weight: 154.25
  • Molecular formula: C10H18O
  • Purity: 98.49%
  • Density: 0.933±0.06 g/cm3
  • For research use only
  • Extracted from natural sources

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.