Learn More
Apexbio Technology LLC Terpinen-4-ol 562-74-3 100mg
SDP
Catalog No. 5000089212
Encompass_Preferred
Terpinen-4-ol (CAS 562-74-3) is a monoterpene alcohol predominantly found in tea tree oil It exhibits broad-spectrum antimicrobial and anti-inflammatory activities attributed to its ability to disrupt microbial cell membranes and modulate pro-inflammatory mediator expression Terpinen-4-ol has demonstrated acaricidal activity particularly relevant to the ocular parasite Demodex spp Its therapeutic potential is being evaluated in clinical studies including investigations for the treatment of Demodex blepharitis This compound is of interest for research focused on antimicrobial mechanisms and host-pathogen interactions
Catalog No. 50-000-89212 Supplier Apexbio Technology LLC Supplier No. C6423100
Terpinen-4-ol (CAS 562-74-3) is a monoterpene alcohol predominantly found in tea tree oil It exhibits broad-spectrum antimicrobial and anti-inflammatory activities attributed to its ability to disrupt microbial cell membranes and modulate pro-inflammatory mediator expression Terpinen-4-ol has demonstrated acaricidal activity particularly relevant to the ocular parasite Demodex spp Its therapeutic potential is being evaluated in clinical studies including investigations for the treatment of Demodex blepharitis This compound is of interest for research focused on antimicrobial mechanisms and host-pathogen interactions