Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terpinen-4-ol | 562-74-3 | MFCD00001562 | 98.6% | 154.25 g/mol | C10H18O | 1 ML
SDP

Catalog No. 5000299910
Encompass_Preferred

Terpinen-4-ol is a naturally occurring monoterpene alcohol (CAS 562-74-3) provided as a 10 mM solution in a 1 mL vial for laboratory research. It is used as a reagent or reference standard in studies of antimicrobial, antifungal, and anti-inflammatory activity, and the supplier Certificate of Analysis reports high purity and standard physicochemical identifiers.

  • Naturally occurring monoterpene alcohol with antimicrobial and anti-inflammatory activity.
  • Supplied as a 10 mM solution in a 1 mL vial.
  • High purity (98.6% GC) suitable for analytical and biological assays.
  • Molecular formula C10H18O; molecular weight 154.25 g/mol.
  • Suitable as a reagent or reference standard in pharmacology and biochemical research.
  • Storage stability for solutions: -80°C (6 months), -20°C (1 month); protect from light.

Catalog No. 50-002-99910 Supplier Medchemexpress LLC Supplier No. HYW01731610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terpinen-4-ol is a naturally occurring monoterpene alcohol (CAS 562-74-3) provided as a 10 mM solution in a 1 mL vial for laboratory research. It is used as a reagent or reference standard in studies of antimicrobial, antifungal, and anti-inflammatory activity, and the supplier Certificate of Analysis reports high purity and standard physicochemical identifiers.

  • Naturally occurring monoterpene alcohol with antimicrobial and anti-inflammatory activity.
  • Supplied as a 10 mM solution in a 1 mL vial.
  • High purity (98.6% GC) suitable for analytical and biological assays.
  • Molecular formula C10H18O; molecular weight 154.25 g/mol.
  • Suitable as a reagent or reference standard in pharmacology and biochemical research.
  • Storage stability for solutions: -80°C (6 months), -20°C (1 month); protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.