Learn More
Medchemexpress LLC Terpinen-4-ol | 562-74-3 | MFCD00001562 | 98.7% | 154.25 g·mol⁻¹ | C10H18O | 10 MG
SDP
Catalog No. 5000299775
Encompass_Preferred
Terpinen-4-ol is an analytical standard of the naturally occurring monoterpene 4-carvomenthenol used for research and analytical applications. It functions as a reference material for chromatographic method development, qualitative identification, and quantitative analysis. The compound has formula C10H18O, molecular weight 154.25 g·mol⁻¹, and CAS 562-74-3; batch COA reports purity ≈98.65% (GC).
  • Analytical standard for chromatography and method validation.
  • High purity (~98.7% by GC) for reliable quantitation.
  • Suitable for qualitative identification and reference libraries.
  • Supplied in measured quantities for laboratory use.
  • Applicable to research in antimicrobial, antifungal, and anti-inflammatory studies.
Catalog No. 50-002-99775 Supplier Medchemexpress LLC Supplier No. HYW017316R10MG
Terpinen-4-ol is an analytical standard of the naturally occurring monoterpene 4-carvomenthenol used for research and analytical applications. It functions as a reference material for chromatographic method development, qualitative identification, and quantitative analysis. The compound has formula C10H18O, molecular weight 154.25 g·mol⁻¹, and CAS 562-74-3; batch COA reports purity ≈98.65% (GC).
  • Analytical standard for chromatography and method validation.
  • High purity (~98.7% by GC) for reliable quantitation.
  • Suitable for qualitative identification and reference libraries.
  • Supplied in measured quantities for laboratory use.
  • Applicable to research in antimicrobial, antifungal, and anti-inflammatory studies.