Learn More
Medchemexpress LLC Terpinen-4-ol | 562-74-3 | MFCD00001562 | 98.7% | 154.25 g/mol | C10H18O | 500 MG
SDP
Catalog No. 5000300931
Encompass_Preferred
Terpinen-4-ol is a monoterpene alcohol commonly used in research for its antimicrobial and pharmacological properties. It exhibits bactericidal and antifungal activity and has reported anti-inflammatory and antitumor effects. The compound is provided at high purity and is suitable for in vitro studies and formulation development, with good solubility in DMSO and water.
  • High purity suitable for research use.
  • Monoterpene alcohol with bactericidal and antifungal activity.
  • Reported anti-inflammatory and antitumor properties.
  • Soluble in DMSO and water for easy formulation.
  • Appropriate for in vitro antimicrobial and pharmacology studies.
  • Available in multiple pack sizes to suit laboratory needs.
Catalog No. 50-003-00931 Supplier Medchemexpress LLC Supplier No. HYW017316500MG
Terpinen-4-ol is a monoterpene alcohol commonly used in research for its antimicrobial and pharmacological properties. It exhibits bactericidal and antifungal activity and has reported anti-inflammatory and antitumor effects. The compound is provided at high purity and is suitable for in vitro studies and formulation development, with good solubility in DMSO and water.
  • High purity suitable for research use.
  • Monoterpene alcohol with bactericidal and antifungal activity.
  • Reported anti-inflammatory and antitumor properties.
  • Soluble in DMSO and water for easy formulation.
  • Appropriate for in vitro antimicrobial and pharmacology studies.
  • Available in multiple pack sizes to suit laboratory needs.