Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Terpinen-4-ol 95%, Thermo Scientific™

Catalog No. AC203750250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC203750250 25 g
AC203751000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC203750250 Supplier Thermo Scientific Chemicals Supplier No. 203750250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2438-10-0
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
InChI Key WRYLYDPHFGVWKC-SNVBAGLBSA-N
Synonym +-terpinen-4-ol, unii-hzk34ln80n, s-p-menth-1-en-4-ol, hzk34ln80n, s-1-isopropyl-4-methyl-3-cyclohexen-1-ol, 4-terpinenol, d, +-4-terpineol, 4-terpineol, +, 1s-1-isopropyl-4-methylcyclohex-3-en-1-ol, 4-carvomenthenol, +
PubChem CID 2724161
IUPAC Name (1S)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
SMILES CC1=CCC(CC1)(C(C)C)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.932
Infrared Spectrum Authentic
Refractive Index 1.4760 to 1.4820 (20°C, 589nm)
Quantity 25 g
Beilstein 06, IV, 250
Specific Gravity 0.929 to 0.935 (20°/20°C)
Specific Rotation +27° to +32° (20°C, 589nm) (neat)
Formula Weight 154.25
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material (+)-Terpinen-4-ol, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.