Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Terpinen-4-ol, 97%, sum of enantiomers

Catalog No. p-194762
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270430250 25 g Glass bottle
AC270431000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270430250 Supplier Thermo Scientific Chemicals Supplier No. 270430250
Only null left
Add to Cart

CAS: 20126-76-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20126-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167108
InChI Key WRYLYDPHFGVWKC-JTQLQIEISA-N
Synonym --terpinen-4-ol, --4-terpineol, unii-8vi196vs5t, p-menth-1-en-4-ol, r--, r-p-menth-1-en-4-ol, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, r, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, 1r, r-1-isopropyl-4-methyl-3-cyclohexen-1-ol, l-4-terpineneol, l-4-terpineol
PubChem CID 5325830
IUPAC Name (1R)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
SMILES CC1=CCC(CC1)(C(C)C)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 212.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (sum of enantiomers) (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4820
Quantity 25 g
Beilstein 06, IV, 250
Specific Gravity 0.933
Specific Rotation Condition −25° (20°C c=neat)
Specific Rotation − 25.00
Solubility Information Solubility in water: very slightly soluble., soluble in alcohol
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (-)-Terpinen-4-ol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.