Learn More
Medchemexpress LLC γ-Terpinene | 99-85-4 | MFCD00001537 | 98.1% | 136.23 g/mol | C10H16 | 500 MG
SDP
Catalog No. 5000301790
Encompass_Preferred
γ-Terpinene is a monoterpene antioxidant used in research to scavenge free radicals and to investigate antimicrobial and pharmacological effects. It is a colorless to light-yellow liquid with formula C10H16 and molecular weight 136.23 g/mol. The compound is supplied at research grade purity and is suitable for formulation in DMSO or common vehicle systems for in vitro and in vivo studies.
  • High purity (98.14%) suitable for analytical and biological assays.
  • Soluble in DMSO at 200 mg/mL for in vitro applications.
  • Colorless to light yellow liquid that is easy to handle and formulate.
  • Stable under recommended storage (pure form: -20°C or 4°C) for extended periods.
  • Compatible with common in vivo vehicle systems for dosing studies.
Catalog No. 50-003-01790 Supplier Medchemexpress LLC Supplier No. HYW020183500MG
γ-Terpinene is a monoterpene antioxidant used in research to scavenge free radicals and to investigate antimicrobial and pharmacological effects. It is a colorless to light-yellow liquid with formula C10H16 and molecular weight 136.23 g/mol. The compound is supplied at research grade purity and is suitable for formulation in DMSO or common vehicle systems for in vitro and in vivo studies.
  • High purity (98.14%) suitable for analytical and biological assays.
  • Soluble in DMSO at 200 mg/mL for in vitro applications.
  • Colorless to light yellow liquid that is easy to handle and formulate.
  • Stable under recommended storage (pure form: -20°C or 4°C) for extended periods.
  • Compatible with common in vivo vehicle systems for dosing studies.