Learn More
Medchemexpress LLC α-terpinene | 99-86-5 | MFCD00001534 | 95.0% | 136.23 g/mol | C10H16 | 1 G
SDP
Catalog No. 5000299482
Encompass_Preferred
α-Terpinene is a monoterpene (C10H16, 136.23 g/mol) found in essential oils of various aromatic plants. It is used as a research reagent and analytical reference and has reported antioxidant, antifungal, and antiparasitic activity.
  • Monoterpene hydrocarbon with formula C10H16.
  • Provided as a laboratory research reagent and analytical standard.
  • Reported antioxidant and antifungal properties.
  • Used in studies of antiparasitic activity and natural product research.
  • Available in small-scale laboratory packaging for analytical use.
Catalog No. 50-002-99482 Supplier Medchemexpress LLC Supplier No. HYW0201821G
α-Terpinene is a monoterpene (C10H16, 136.23 g/mol) found in essential oils of various aromatic plants. It is used as a research reagent and analytical reference and has reported antioxidant, antifungal, and antiparasitic activity.
  • Monoterpene hydrocarbon with formula C10H16.
  • Provided as a laboratory research reagent and analytical standard.
  • Reported antioxidant and antifungal properties.
  • Used in studies of antiparasitic activity and natural product research.
  • Available in small-scale laboratory packaging for analytical use.