Learn More
Medchemexpress LLC (-)-α-Terpineol | 10482-56-1 | MFCD00075926 | 154.25 | 10 G
SDP
Catalog No. 5000415662
Encompass_Preferred
(-)-α-Terpineol is a monoterpene compound derived from Melaleuca alternifolia, known for its antioxidant and anti-inflammatory properties. Research indicates its potential in modulating TNF-α levels and serum thiobarbituric acid reactive substances, as well as improving insulin sensitivity.
  • Exhibits antioxidant activity
  • Exhibits anti-inflammatory activity
  • Decreases TNF-α levels
  • Reduces serum thiobarbituric acid reactive substances
  • Increases insulin sensitivity
Catalog No. 50-004-15662 Supplier Medchemexpress LLC Supplier No. HYN146710G
(-)-α-Terpineol is a monoterpene compound derived from Melaleuca alternifolia, known for its antioxidant and anti-inflammatory properties. Research indicates its potential in modulating TNF-α levels and serum thiobarbituric acid reactive substances, as well as improving insulin sensitivity.
  • Exhibits antioxidant activity
  • Exhibits anti-inflammatory activity
  • Decreases TNF-α levels
  • Reduces serum thiobarbituric acid reactive substances
  • Increases insulin sensitivity