Learn More
Medchemexpress LLC Terpineol | 8000-41-7 | 250 G
SDP
Catalog No. 5000422030
Encompass_Preferred
Terpineol is a monoterpenoid alcohol compound widely found in plants, particularly in citrus fruit peels. It exhibits antioxidant, anti-inflammatory, and antibacterial activities and is commonly used in the fragrance industry as an additive in food, beverages, cosmetics, and perfumes.
  • Monoterpenoid alcohol compound
  • Found in citrus fruit peels
  • Possesses antioxidant activity
  • Possesses anti-inflammatory activity
  • Possesses antibacterial activity
  • Used as an additive in food
  • Used as an additive in beverages
  • Used as an additive in cosmetics
  • Used as an additive in perfumes
Catalog No. 50-004-22030 Supplier Medchemexpress LLC Supplier No. 250GHYN7851
Terpineol is a monoterpenoid alcohol compound widely found in plants, particularly in citrus fruit peels. It exhibits antioxidant, anti-inflammatory, and antibacterial activities and is commonly used in the fragrance industry as an additive in food, beverages, cosmetics, and perfumes.
  • Monoterpenoid alcohol compound
  • Found in citrus fruit peels
  • Possesses antioxidant activity
  • Possesses anti-inflammatory activity
  • Possesses antibacterial activity
  • Used as an additive in food
  • Used as an additive in beverages
  • Used as an additive in cosmetics
  • Used as an additive in perfumes