Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-α-Terpineol, ≥97.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018186 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8186 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8186 Supplier MilliporeSigma Supelco Supplier No. 830735MLF
Add to Cart
Add to Cart
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 7785-53-7
Density 0.94 g/mL (at 20°C)
Flash Point 90°C
Physical Form Neat
Linear Formula C10H18 O
MDL Number MFCD00171435
Synonym (R)-2-(4-Methyl-3-cyclohexenyl)isopropanol; (R)-p-Menth-1-en-8-ol
Type Analytical Standard
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥97.0% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.