Learn More
Medchemexpress LLC α-terpineol-d3 | 203633-12-9 | 98.1% | 157.27 g/mol | C10H15D3O | 10 MG
SDP
Catalog No. 5000289751
Encompass_Preferred
α-Terpineol-d3 is the deuterium-labeled isotopologue of α-terpineol provided as a stable-isotope reference standard for analytical and research use. It is intended for use as an internal standard and tracer in mass spectrometry and isotope-labeling studies, and is supplied in research-scale quantities for analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for GC-MS and LC-MS quantitative analyses.
  • High chemical purity (~98.1%).
  • Provided in a small package size appropriate for analytical work (10 mg).
  • Useful for tracer and isotope-labeling experiments and method development.
Catalog No. 50-002-89751 Supplier Medchemexpress LLC Supplier No. HYN5142S10MG
α-Terpineol-d3 is the deuterium-labeled isotopologue of α-terpineol provided as a stable-isotope reference standard for analytical and research use. It is intended for use as an internal standard and tracer in mass spectrometry and isotope-labeling studies, and is supplied in research-scale quantities for analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for GC-MS and LC-MS quantitative analyses.
  • High chemical purity (~98.1%).
  • Provided in a small package size appropriate for analytical work (10 mg).
  • Useful for tracer and isotope-labeling experiments and method development.