Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terpineol, Practical, Spectrum™ Chemical
SDP

Catalog No. 18614333 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-333 500 mL Amber Glass Bottle
18-614-329 100 mL Amber Glass Bottle
18-614-332 4 L Amber Glass Bottle
18-614-330 20 L Amber Glass Bottle
18-614-331 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-614-333 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1039500MLGL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8000-41-7
Molecular Weight (g/mol) 154.24 g/mol

Specifications

CAS Max % 1
Boiling Point 213°C to 224°C
Packaging Amber Glass Bottle
Refractive Index 1.482-1.484
Quantity 500 mL
Specific Gravity 0.932 - 0.937
Grade Practical
Physical Form Liquid
Chemical Name or Material Terpineol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.