Learn More
Cayman Chemical (−)-Terpinen-4-ol, Purity: ≥95%, CAS: 20126-76-5, 1 g
SDP
Catalog No. 502917120
Encompass_Preferred
A monoterpene alcohol with arrhythmogenic and anticonvulsant activities; induces extrasystoles in isolated rat left atria at 100 µM; induces supraventricular tachycardia in rats at 10 mg/kg; increases the latency to seizure and decreases the number of seizures in a rat model of PTZ-induced epilepsy
Catalog No. 50-291-7120 Supplier Cayman Chemical Supplier No. 389641 G
A monoterpene alcohol with arrhythmogenic and anticonvulsant activities; induces extrasystoles in isolated rat left atria at 100 µM; induces supraventricular tachycardia in rats at 10 mg/kg; increases the latency to seizure and decreases the number of seizures in a rat model of PTZ-induced epilepsy