Learn More
AdipoGen Terpyridine
SDP
Supplier:  AdipoGen CDXT0113G005
Encompass_Preferred
Chemical. CAS 1148-79-4. Formula C15H11N3. MW 233.3. Synthetic. Reagent for the spectrophotometric det. of Ag and Ru. Tridentate ligand that binds metals at three meridional sites giving two adjacent 5-membered MN2C2 chelate rings. It forms complexes with most transition metal ions. It is also a potent inducer of nitrate and nitric oxide NO synthase iNOS protein and mRNA production in the cultured murine macrophage RAW 264.7 cell line.
Catalog No. 50-256-0003