Learn More
Takara Bio TERRA PCR MIX BLOOD
SDP
Catalog No. NC0221868
Encompass
Direct PCR Kits- With Terra PCR Direct, amplify difficult GC-rich (>70%) DNA effortlessly, even in the presence of PCR inhibitors.
Catalog No. NC0221868 Supplier Takara Bio Supplier No. 639270
Direct PCR Kits- With Terra PCR Direct, amplify difficult GC-rich (>70%) DNA effortlessly, even in the presence of PCR inhibitors.