Learn More
Med Vet International Zoetis Terramycin Ophthalmic Ointment, 3.5g / 1/8oz.
Catalog No. 502828304
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Terramycin (oxytetracycline HCl) Ophthalmic Ointment by Zoetis with Polymyxin B Sulfate is indicated for the prevention and local treatment of superficial ocular infections due to oxytetracycline- and polymyxin-sensitive organisms, including infections due to Streptococci, Rickettsiae, E. coli, and A. aerogenes, such as conjunctivitis, keratitis, pink eye, corneal ulcer, blepharitis in dogs, cats, cattle, sheep, and horses; ocular infections due to secondary bacterial complications of distemper in dogs; and bacterial inflammatory conditions that may occur secondary to other infectious diseases in the above species. NOT FOR SALE IN CALIFORNIA
Catalog No. 50-282-8304 Supplier Med Vet International Supplier No. RXTERRA1/8
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Terramycin (oxytetracycline HCl) Ophthalmic Ointment by Zoetis with Polymyxin B Sulfate is indicated for the prevention and local treatment of superficial ocular infections due to oxytetracycline- and polymyxin-sensitive organisms, including infections due to Streptococci, Rickettsiae, E. coli, and A. aerogenes, such as conjunctivitis, keratitis, pink eye, corneal ulcer, blepharitis in dogs, cats, cattle, sheep, and horses; ocular infections due to secondary bacterial complications of distemper in dogs; and bacterial inflammatory conditions that may occur secondary to other infectious diseases in the above species. NOT FOR SALE IN CALIFORNIA