Learn More
AdipoGen Terreic acid
SDP
Supplier:  AdipoGen BVT0477M005
Encompass_Preferred
Chemical. CAS 121-40-4. Formula C7H6O4. MW 154.1. Isolated from Aspergillus terreus.. Mycotoxin. Antibacterial and anticancer agent. A cell-permeable quinone epoxide antibiotic. Covalent inhibitor of the bacterial enzyme UDP-N-acetylglucosamine 1-carboxyvinyltransferase MurA, which catalyzes the first committed step of bacterial cell wall biosynthesis. More potent than the known MurA inhibitor Fosphomycin. Reversible, substrate competitive and selective inhibitor of Brutons tyrosine kinase BTK catalytic activity, an important factor in B-cell and mast cell activation. Shown to bind to the BTK pleckstrin homology domain BTK-PH and block the interaction between the BTK-PH and PKC, affecting the catalytic activity of BTK but not of PKC. Has minimal effect on Lyn, Syk, PKA, casein kinase I, ERK1, ERK2, and p38 kinase activities. Antioxidant.
Catalog No. 50-256-1882