Learn More
AdipoGen Terrein
SDP
Supplier:  AdipoGen BVT0193M005
Encompass_Preferred
Chemical. CAS 582-46-7. Formula C8H10O3. MW 154.2. Isolated from Aspergillus terreus. Antibacterial. Phytotoxin. Melanogenesis inhibitor by reducing tyrosinase production via extracellular signal-regulated protein kinase ERK activation. Keratinocyte and epidermis proliferation inhibitor. Mycotoxin. Anti-inflammatory modulator in pulpal inflammation. Angiogenesis inhibitor through angiogenin secretion inhibition. Collagen-induced platelet aggregation inhibitor. Antioxidant agent that enhances osseointegration by decreasing the level of ROS and having a potentially synergistic effect on osteoblast differentiation. Proteasome inhibitor.
Catalog No. 50-256-2072