Learn More
Bioworld Terrestrial (Cypripedium) Orchid Medium, 25 L
Supplier:  Bioworld 306301943
Encompass_Preferred
Terrestrial (Cypripedium) Orchid MediumContains the Macronutrients, Micronutrients, Glucose, and Agar as described by Steele (1996) with the addition of Calcium Nitrate and CaseinSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 27.44 g/LpH: 5.0–6.0Partially soluble in water
Catalog No. 50-254-984