Learn More
TARGETMOL CHEMICALS INC Terrestrosin D 10MG
SDP
Catalog No. 502876547
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Terrestrosin D can induce apoptotic cell death and inhibit angiogenesis in xenograft tumor cells, cell cycle arrest and induction of apoptosis in cancer cells and endothelial cells might be plausible mechanisms of actions for the observed antitumor and antiangiogenic activities of terrestrosin D. Purity 99.54%
Catalog No. 50-287-6547 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S088610MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Terrestrosin D can induce apoptotic cell death and inhibit angiogenesis in xenograft tumor cells, cell cycle arrest and induction of apoptosis in cancer cells and endothelial cells might be plausible mechanisms of actions for the observed antitumor and antiangiogenic activities of terrestrosin D. Purity 99.54%