Learn More
Medchemexpress LLC Terrestrosin D | 179464-23-4 | 5 MG
SDP
Catalog No. 5000419593
Encompass_Preferred
Terrestrosin D is an orally active apoptosis inducer that causes cell cycle arrest at G1 and S phases, decreases mitochondrial membrane potential, and inhibits the growth of cancer cells and endothelial cells. It is being studied for potential use in castration-resistant prostate cancer and pulmonary fibrosis.
  • Induces cell cycle arrest at G1 and S phases.
  • Reduces mitochondrial membrane potential.
  • Inhibits the growth of cancer cells and endothelial cells.
  • Exhibits cytotoxicity in human normal liver and embryonic kidney cells.
  • Potently inhibits the growth of human prostate cancer cell lines.
  • Induces caspase-independent apoptosis in prostate cancer and endothelial cells.
  • Leads to mitochondrial membrane potential depolarization.
  • Increases VEGF secretion.
  • Suppresses xenograft tumor growth in mice.
  • Attenuates bleomycin-induced pulmonary inflammation and fibrosis in mice.
Catalog No. 50-004-19593 Supplier Medchemexpress LLC Supplier No. HYN50745MG
Terrestrosin D is an orally active apoptosis inducer that causes cell cycle arrest at G1 and S phases, decreases mitochondrial membrane potential, and inhibits the growth of cancer cells and endothelial cells. It is being studied for potential use in castration-resistant prostate cancer and pulmonary fibrosis.
  • Induces cell cycle arrest at G1 and S phases.
  • Reduces mitochondrial membrane potential.
  • Inhibits the growth of cancer cells and endothelial cells.
  • Exhibits cytotoxicity in human normal liver and embryonic kidney cells.
  • Potently inhibits the growth of human prostate cancer cell lines.
  • Induces caspase-independent apoptosis in prostate cancer and endothelial cells.
  • Leads to mitochondrial membrane potential depolarization.
  • Increases VEGF secretion.
  • Suppresses xenograft tumor growth in mice.
  • Attenuates bleomycin-induced pulmonary inflammation and fibrosis in mice.