Learn More
Medchemexpress LLC Terrestrosin D | 179464-23-4 | 99.3% | 1049.16 g·mol⁻¹ | C50H80O23 | 10 MG
SDP
Catalog No. 5000289808
Encompass_Preferred
Terrestrosin D is a steroidal saponin isolated from Tribulus terrestris L., used in research to induce cell cycle arrest and apoptosis in cancer cells and to inhibit angiogenesis. It is supplied at high purity for in vitro biochemical and cellular assays.
  • Steroidal saponin with reported pro-apoptotic activity
  • Demonstrated ability to induce cell cycle arrest
  • Shows antiangiogenic effects in experimental models
  • High purity suitable for in vitro research and screening
  • Available as small research pack sizes and solution options for assay flexibility
Catalog No. 50-002-89808 Supplier Medchemexpress LLC Supplier No. HYN507410MG
Terrestrosin D is a steroidal saponin isolated from Tribulus terrestris L., used in research to induce cell cycle arrest and apoptosis in cancer cells and to inhibit angiogenesis. It is supplied at high purity for in vitro biochemical and cellular assays.
  • Steroidal saponin with reported pro-apoptotic activity
  • Demonstrated ability to induce cell cycle arrest
  • Shows antiangiogenic effects in experimental models
  • High purity suitable for in vitro research and screening
  • Available as small research pack sizes and solution options for assay flexibility