Learn More
Sigma Aldrich Fine Chemicals Biosciences Terrfc Broth EZMxTM pow500ML
Catalog No. 502901783
Encompass
Terrific Broth is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains glycerol peptides amino acids water-soluble vitamins and carbohydrates.
Catalog No. 50-290-1783 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T91796X500ML
Terrific Broth is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains glycerol peptides amino acids water-soluble vitamins and carbohydrates.