Learn More
AG Scientific Inc Terrific Broth 1 KG
Catalog No. 502268402
Encompass_Preferred
Terrific Broth is an enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant protein. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.pH (4.76% solution at 25ºC): 6.8-7.4
Catalog No. 50-226-8402 Supplier AG Scientific Inc Supplier No. T27051KG
Terrific Broth is an enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant protein. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.pH (4.76% solution at 25ºC): 6.8-7.4