Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Terrific Broth, Case 10 X 1000 mL
SDP

Catalog No. 507517391
Encompass_Preferred

Terrific Broth was developed by Tartoff and Hobbs to increase yields in plasmid bearing E. coli. It is an enriched medium suitable to increase the yield of plasmid DNA derived from E. coli strains, for non-selective cultivation of E. coli strains (for cloning high yield DNA plasmid production, and high yield production of recombinant proteins), and selective cultivation when appropriate antibiotics are added. The large amount of tryptone and yeast extract in the medium allows the bacteria to produce a higher plasmid yield.

Autoclave sterilized
Ready to use

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7391 Supplier Quality Biological Inc Supplier No. 340071131CS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terrific Broth was developed by Tartoff and Hobbs to increase yields in plasmid bearing E. coli. It is an enriched medium suitable to increase the yield of plasmid DNA derived from E. coli strains, for non-selective cultivation of E. coli strains (for cloning high yield DNA plasmid production, and high yield production of recombinant proteins), and selective cultivation when appropriate antibiotics are added. The large amount of tryptone and yeast extract in the medium allows the bacteria to produce a higher plasmid yield.

Autoclave sterilized
Ready to use

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.