Learn More
Bioworld Terrific Broth, 24-gram capsules, 200 Capsule(s)
Catalog No. 50254376
Encompass_Preferred
Terrific BrothUsed with glycerol in cultivating recombinant strains of Escherichia coliConvenient premeasured and pre-mixed 24g capsulesHighly enriched medium developed by Tartoff and Hobbs to improve yield in plasmid-bearing E. colipH: 7.2Dissolve in 2 capsules in 1L purified water with 4mL glycerolPrepared Appearance: light to medium amber brothSoluble in water/glycerol mixtureContains:Pancreatic Digest of CaseinYeast ExtractDipotassium PhosphateMonopotassium Phosphate
Catalog No. 50-254-376 Supplier Bioworld Supplier No. 306200823
Terrific BrothUsed with glycerol in cultivating recombinant strains of Escherichia coliConvenient premeasured and pre-mixed 24g capsulesHighly enriched medium developed by Tartoff and Hobbs to improve yield in plasmid-bearing E. colipH: 7.2Dissolve in 2 capsules in 1L purified water with 4mL glycerolPrepared Appearance: light to medium amber brothSoluble in water/glycerol mixtureContains:Pancreatic Digest of CaseinYeast ExtractDipotassium PhosphateMonopotassium Phosphate