Learn More
AG Scientific Inc Terrific Broth 500 G
Catalog No. 502268403
Encompass_Preferred
Terrific Broth is an enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant protein. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.pH (4.76% solution at 25ºC): 6.8-7.4
Catalog No. 50-226-8403 Supplier AG Scientific Inc Supplier No. T2705500GM
Terrific Broth is an enriched media for maintenance and propagation of E. Coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant protein. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.pH (4.76% solution at 25ºC): 6.8-7.4