Learn More
Teknova Terrific Broth. 900mL, Sterile. (P7100 10X Phosphate Buffer Included)
SDP
Catalog No. 50843343
Encompass
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.
This item is not returnable. View return policy
Catalog No. 50-843-343 Supplier Teknova Supplier No. T7000
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.