Learn More
Teknova Terrific Broth Agar Plates with Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387725
Encompass
1.20% Tryptone, 2.40% Yeast Extract, 1.60% Glycerol, 1.25% Potassium Phosphate Dibasic, 0.23% Potassium Phosphate Monobasic, 1.50% Agar, 50g/mL Kanamycin
Catalog No. 50-238-7725 Supplier Teknova Supplier No. T49504PK
1.20% Tryptone, 2.40% Yeast Extract, 1.60% Glycerol, 1.25% Potassium Phosphate Dibasic, 0.23% Potassium Phosphate Monobasic, 1.50% Agar, 50g/mL Kanamycin