Learn More
Teknova Terrific Broth Agar Plates with Carbenicillin-100. 150mm Plates, Sterile. 8 pack.
SDP
Catalog No. 502387310
Encompass
1.2% Tryptone, 2.4% Yeast Extract, 1.6% Glycerol, 2.0% Agar, 72mM Potassium Phosphate Dibasic,17mM Potassium Phosphate Monobasic, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7310 Supplier Teknova Supplier No. T49788PK
1.2% Tryptone, 2.4% Yeast Extract, 1.6% Glycerol, 2.0% Agar, 72mM Potassium Phosphate Dibasic,17mM Potassium Phosphate Monobasic, 100g/mL Carbenicillin Disodium Salt