Learn More
Teknova Terrific Broth Agar with 1.6% Glycerol. 1kg
SDP
Catalog No. 502388188
Encompass
1.2% Tryptone, 2.4% Yeast Extract, 1.5% Agar, 72mM Potassium Phosphate, Dibasic, 17mM Potassium Phosphate, Monobasic, 1.6% Glycerol
Catalog No. 50-238-8188 Supplier Teknova Supplier No. T0385
1.2% Tryptone, 2.4% Yeast Extract, 1.5% Agar, 72mM Potassium Phosphate, Dibasic, 17mM Potassium Phosphate, Monobasic, 1.6% Glycerol