Learn More
Boca Scientific Inc Terrific Broth, Auto-Induction Medium, 500g
Catalog No. NC1894479
Encompass
Terrific Broth (Auto-Induction Medium) is a dehydrated powder for the preparation of Terrific Broth without trace elements. This medium is supplemented with glucose and alpha lactose for the auto induction of protein expression under the control of IPTG-inducible promoters in E. coli. The Auto-Induction Medium (AIM) provides an efficient means of protein expression that requires far less attention by the user than conventional IPTG induction. With the Auto-Induction Medium, it is no longer necessary to monitor cell density and to add IPTG at the proper stage, as the medium contains an optimized ratio of glucose and alpha lactose as carbon sources. This system offers unmatched convenience and simplifies manual or automatic induction and analysis of multiple clones compared to conventional IPTG induction. One 500g bottle prepares 9L.
Catalog No. NC1894479 Supplier Boca Scientific Inc Supplier No. GCM19.0500
Terrific Broth (Auto-Induction Medium) is a dehydrated powder for the preparation of Terrific Broth without trace elements. This medium is supplemented with glucose and alpha lactose for the auto induction of protein expression under the control of IPTG-inducible promoters in E. coli. The Auto-Induction Medium (AIM) provides an efficient means of protein expression that requires far less attention by the user than conventional IPTG induction. With the Auto-Induction Medium, it is no longer necessary to monitor cell density and to add IPTG at the proper stage, as the medium contains an optimized ratio of glucose and alpha lactose as carbon sources. This system offers unmatched convenience and simplifies manual or automatic induction and analysis of multiple clones compared to conventional IPTG induction. One 500g bottle prepares 9L.