Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Teknova Terrific Broth Complete 1L GMP
SDP

Catalog No. 5001082685
Encompass

Terrific Broth Complete with Animal-Free Soytone 1000mL Sterile

Catalog No. 50-010-82685 Supplier Teknova Supplier No. 235188PK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terrific Broth Complete with Animal-Free Soytone 1000mL Sterile

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.