Learn More
Teknova Terrific Broth Complete, 2L, Sterile.
SDP
Catalog No. NC1646267
Encompass
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.
This item is not returnable. View return policy
Catalog No. NC1646267 Supplier Teknova Supplier No. T8060
Terrific broth is a rich, nutritionally complete liquid growth media that is widely used in molecular biology and microbiology laboratories. Its high protein and carbohydrate content results in high bacterial and plasmid yields, making it an ideal media for DNA isolation and other molecular techniques.